42736.K2
42736.K2EA
78.9
EUR
InStock
42736.K2
Potassium hydroxyde solution standardisée 1.0 N dans le methanol
Potassium hydroxyde
CAS No.: 1310-58-3
Molecular Formula: KOH
Formula Weight: 56.11
Physical Form: Liquid
Appearance: Colourless
Odour: Like alcohol
UN No.: 3286
MDL No.: MFCD00003553