42865.06
42865.06EA
346
EUR
InStock
42865.06
Indium (I) bromure ≥99.999% (base métaux), Puratronic®
Indium (I) bromure
Purity: 99.999%
CAS No.: 14280-53-6
Molecular Formula: InBr
Formula Weight: 194.73
Physical Form: Polycrystalline Lump
Density: 4.96
Melting Point: 220°
Boiling Point: 662°
UN No.: 3260
MDL No.: MFCD00064630