Organic reference standard, Theobromine, analytical standard, Supelco®
Fournisseur: Merck
42993-100MG
42993-100MGEA 50.5 EUR InStock
42993-100MG
Organic reference standard, Theobromine, analytical standard, Supelco®
Étalons Étalons organiques
Theobromine is an active bronchodilator drug ingredient used in the treatment of acute and chronic asthma. It is a naturally present alkaloid in a wide variety of foods and beverages, such as coffee, cocoa, tea, yerba mate and energizing drinks.
Phosphosdiesterase inhibitor, weak adenosine receptor antagonist; diuretic, smooth muscle relaxant.
Theobromine may be used as a reference standard for the determination of theobromine metabolites in human plasma and urine after consumption of soluble cocoa products with different methylxanthine contents by high-performance liquid chromatography and diode array detection (HPLC-DAD) method.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e276ff72e4dfe7169d0722d3fecb94b19745f7b3&vid=3a15da90235768e2aa7c775f7360cefbc3d00885&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité