43058.P5
43058.TD
43058.P5EA
0
EUR
InStock
43058.P5
43058.TD
Calcium sulfate, Drierite™ 10-20 mesh pour séchage
Calcium sulphate
CAS No.: 7778-18-9
Physical Form: Granular, -10+20 Mesh
Density: 2.96
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Merck Reference: 14,1706
MDL No.: MFCD00010912