Synonyms:
Methylbenzene, Phenylmethane, FM approval
43061.K2
43061.K2EA
54.1
EUR
InStock
43061.K2
Toluène ≥99.8%, Qualité environnementale
Toluène
Purity: 99.8+%
CAS No.: 108-88-3
Molecular Formula: C6H5CH3
Formula Weight: 92.14
Flash Point: 4°(40°F)
Physical Form: Liquid
Density: 0.867
Refractive Index: 1.4967
Odour: Aromatic
Melting Point: -93°
Boiling Point: 111°
Beilstein Reference: 635760
Merck Reference: 14,9529
UN No.: 1294
MDL No.: MFCD00008512
Suitable for pesticide residue analysis