43082.K2
43082.K2EA
47.7
EUR
InStock
43082.K2
Acide sulfurique solution standardisée 3.0 N en solution aqueuse
Acide sulfurique
CAS No.: 7664-93-9
Molecular Formula: H2O4S
Formula Weight: 98.08
Physical Form: Liquid
Appearance: Colourless to pale yellow
Odour: Acrid
UN No.: 2796
MDL No.: MFCD00064589