43142.AD
43142.AK
43142.ADEA
41.4
EUR
InStock
43142.AD
43142.AK
2,3-Diméthylbutane ≥98%
2,3-Diméthylbutane
Purity: 98+%
CAS No.: 79-29-8
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -29°(-20°F)
Physical Form: Liquid
Density: 0.662
Refractive Index: 1.375
Appearance: Colourless
Odour: Mild
Melting Point: -129°
Boiling Point: 57-59°
Beilstein Reference: 1730737
UN No.: 2457
MDL No.: MFCD00008925