Synonyms:
Neodymium triflate, Neodymium(III) trifluoromethanesulphonate
43393.09
43393.09EA
102
EUR
InStock
43393.09
Trifluoromethanesulfonate de néodyme (III) 98%, REacton®
Trifluoromethanesulfonate de néodyme (III)
Purity: 98%
CAS No.: 34622-08-7
Molecular Formula: Nd(OSO2CF3)3
Formula Weight: 591.44
Physical Form: Powder
Density: 1.7
MDL No.: MFCD00192343