43476.YK
43476.YKEA
138
EUR
InStock
43476.YK
Titane ≥99.8% (base métaux), feuille fine, Thickness: 0.0127 mm (0.0005 in)
Titane
Purity: 99.8%
CAS No.: 7440-32-6
Physical Form: ≈0.46g/90x90mm, Not certified pinhole free
Odour: Odourless
MDL No.: MFCD00011264