Synonyms:
Saline, NaCl, Sea salt, Halite
43631.19
43631.19EA
193
EUR
InStock
43631.19
Sodium chlorure pour la spectroscopie
Sodium chlorure
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: Powder
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477