Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 100 ppm
43972.AP
43972.APEA
95.7
EUR
InStock
43972.AP
5-Ethylidene-2-norbornene (mélange endo + exo) ≥98% stabilisé
5-Ethylidene-2-norbornene
Stab. with 100ppm 4-tert-butylcatechol
Purity: 98%
CAS No.: 16219-75-3
Molecular Formula: C9H12
Formula Weight: 120.19
Flash Point: 38°(101°F)
Physical Form: Liquid
Density: 0.89
Boiling Point: 146°
UN No.: UN3295
MDL No.: MFCD00167576