Synonyms:
Gadolinium (III) oxide
44020.14
44020.14EA
185
EUR
InStock
44020.14
Trioxyde de digadolinium ≥99.99% (REO, base d'oxyde de terre rare), nanopoudre
Trioxyde de digadolinium
Purity: 99.99+%
CAS No.: 12064-62-9
Molecular Formula: Gd2O3
Formula Weight: 362.50
Physical Form: 20-40nm Powder
Density: 7.407
Melting Point: 2330°
Merck Reference: 14,4326
MDL No.: MFCD00011026