Synonyms:
Yttrium oxide, Yttrium (III) oxide
44048.14
44048.14EA
152
EUR
InStock
44048.14
Yttrium (III) oxyde ≥99.995% (REO, base d'oxyde de terre rare), nanopoudre, REacton®
Yttrium (III) oxyde
Purity: 99.995%
CAS No.: 1314-36-9
Molecular Formula: Y2O3
Formula Weight: 225.81
Physical Form: 25-50nm APS Powder
Density: 5.01
Appearance: White
Odour: Odourless
Melting Point: 2410°
Merck Reference: 14,10107
MDL No.: MFCD00011473