44177.MQ
44177.MQEA
129
EUR
InStock
44177.MQ
Tin plating solution, sans électricité part B
Tin plating solution
Physical Form: Liquid
Appearance: Colourless
Odour: Acrid
UN No.: UN1789
MDL No.: MFCD03703331
Applications: To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection