ampouled under argon
44182.14
44182.14EA
516
EUR
InStock
44182.14
Lanthane ≥99.7% (REO, base d'oxyde de terre rare), en poudre -40 mesh
Lanthane
Purity: 99.9%
CAS No.: 7439-91-0
Physical Form: Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Gray to silver
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00011066