Magnetic stirrer, myPlate
Fournisseur: IKA
25005156
442-1262EA 414 EUR InStock
442-1262
Magnetic stirrer, myPlate
Agitateurs Agitateurs magnétiques
The myPlate magnetic stirrer has similar technical features to the popular colour squid and big squid magnetic stirrers together with the option of creative individualisation. The interchangeable personal frame allows you to design the set-up plate of the myPlate by yourself.
  • High chemical resistance
  • Electronically controlled motor
  • Interchangeable frame for customisation
  • Proven high functionality
  • Digital speed indicator
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
The stirrer is supplied with a decorative image template, that can easily be exchanged through the convenient interchangeable frame. Some people have family photos on their desk, you can convert your magnetic stirrer into a canvas displaying your favorite pictures.
Certifications: IP protection class according to DIN EN 60529: IP 54
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité