VWR®, Advanced hotplates
Fournisseur: VWR Collection
442-1269EA 815 EUR InStock
442-1269
VWR®, Advanced hotplates
Appareils chauffants Plaques chauffantes
VWR® Advanced hotplates are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With temperature ranges up to 500 °C VWR hotplates provide superior temperature control.
  • Excellent temperature uniformity
  • Easy to read, large LCD displays temperatures
  • Industry-leading safety features
  • Cool touch and chemical resistant housing
  • 5 year warranty
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays temperature settings and intuitive icons.
Reflective, white ceramic top is chemical-resistant and easy to clean. A built-in memory recall provides users with the last set temperature, even after the unit has been turned off. For additional safety, a large, hot symbol warning light is illuminated when the hot plate is in use, this remains lit until the top plate is below 40 °C. Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
The low profile design utilises minimal bench space and allows use within a fume hood. Unit housing is constructed of a heat and chemical resistant polymer that remains cool to the touch during operation. The spill resistant design channels fluids away from internal components in the event of a spill.
Corresponding magnetic hotplate stirrers (heating and stirring 442-1265, 442-1268, 442-1270 and 442-1274) or magnetic stirrers (stirring only 442-1310) are also available.
Certifications: RoHS compliant. TUV listed.
Information de livraison: Units are supplied with a 234 cm three-wire cord and plug.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité