VWR®, Advanced Magnetic Stirrers
Fournisseur: VWR Collection
442-1310EA 741 EUR InStock
442-1310
VWR®, Advanced Magnetic Stirrers
Agitateurs Agitateurs magnétiques
VWR® Advanced magnetic stirrers are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With stirring speeds reaching 1600 rpm, VWR® magnetic stirrers provide proper mixing. Available with resin plate.
  • Excellent stirring
  • Easy to read, large LCD displays speed
  • Cool touch and chemical resistant housing
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays speed settings and intuitive icons.
Magnetic stirrers are available with resin topMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité