Cimarec™ i 20C Controller, 100 to 240 V
Fournisseur: H + P LABORTECHNIK
50090773
442-3141EA 863 EUR InStock
442-3141
Cimarec™ i 20C Controller, 100 to 240 V
Agitateurs Outils d'agitation
Simplify operations with the Thermo Scientific™ Cimarec™ i 20C Controller, a user-friendly, menu-driven microprocessor controller that is included with the Micro, Mini, Compact, Maxi and Telesystems stirrers.
  • Single-handed operation with straightforward turn and press adjustment wheel
  • Three individual program, store and recall keys for rapid start-up
  • Last used settings stored in memory
  • Rocking/shake mode reverses stir bar direction for a gentle back and forth stirring motion
  • Four power settings to decrease power consumption and lower heat output
Informations de commande: Order as replacement controller only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité