44243.6N
44243.6NEA
141
EUR
InStock
44243.6N
Titane ≥99.6% (base métaux), feuille fine, Thickness: 0.0127 mm (0.0005 in)
Titane
Purity: 99.6%
CAS No.: 7440-32-6
Molecular Formula: Commercial Pure Grade 4
Physical Form: ≈0.515g/90x100mm, Light Tight
Odour: Odourless