44325.18
44325.18EA
542
EUR
InStock
44325.18
Gallium ≥99.99999% (base métaux), en pastilles, Ø 6 mm
Gallium
Purity: 99.99999%
CAS No.: 7440-55-3
Molecular Formula: Requires overnight shipping with cold pack
Storage Temperature: 2°C to 8°C
Physical Form: max. weight 0.7g per pellet
Appearance: Silver grey
UN No.: 2803
MDL No.: MFCD00134045