VWR®, Multi Purpose Clamps, 3-Prong
Fournisseur: VWR Collection
444-0916EA 65.2 EUR InStock
444-0916
VWR®, Multi Purpose Clamps, 3-Prong
Pinces Pinces à usage général
The multi purpose clamp is designed to securely hold every type of laboratory glassware and apparatus.
  • Large grip adjustment range
  • Three prong construction
  • Dual adjust
Long, seamless nickel-plated brass tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms, which allow the clamps to be rotated 360°.
Extension arms also allow placement of apparatus at various distances from lab-frames without compromising the integrity of the experiment.
Informations de commande: An additional holder must be purchased in order to attach clamps to frames or other apparatus. Supplied with non-slip vinyl sleeves and, for temperatures above 100 °C, fiberglass covers.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité