Orbital shaker, Mini-100 Orbital-Genie™
Fournisseur: SCIENTIFIC INDUSTRIES ELMIS
SI-M102
444-1075EA 1290 EUR InStock
444-1075
Orbital shaker, Mini-100 Orbital-Genie™
Agitateurs Agitateurs orbitaux
Mini-100 Orbital-Genie™ is ideal for gentle shaking applications, with variable shaking speed up to 100 min⁻¹.
  • Non-slip, rubber-coated platform with spill-proof edges
  • Ideal for odd shaped vessels and containers
  • No additional accessories required
  • Optional stacking tray increases capacity
Features variable shaking speed, which allows user to create any mixing action from a smooth vortex to sloshing aeration or anything in between. The combination of weight and specially designed feet prevent walking even at highest speed.
Informations de commande: Optional stacking trays are available to increase the capacity of the shaker, up to 2 trays can be stacked and no tools are required to fix the trays in place. Please order accessories separately.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité