Thermoshaker, Matrix Orbital
Fournisseur: IKA
30000627
444-1277EA 1874 EUR InStock
444-1277
Thermoshaker, Matrix Orbital
Agitateurs Agitateurs incubateurs
The IKA Matrix Orbital is part of a powerful product family of thermoshakers which have numerous user-friendly features.
  • Large, clear display with a convenient menu structure allowing intuitive operation
  • Speeds of up to 3000 rpm and shaking diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for optimal mixing
  • RS232 and USB interfaces
IKA Matrix Orbital mixes and is suitable foruse with variable inserts. Whether it is blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications. No cross-contamination and an optimal mixing result.
Certifications: Meets ASTM standards. EN-342. IP Protection Class according to DIN EN 60529: IP 21
Information de livraison: Supplied with inserts for 0,2 ml PCR vials and 0,5 and 1,5/2,0 ml reaction vials.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité