MultiPlate Genie® Microplate Shaker
Fournisseur: SCIENTIFIC INDUSTRIES ELMIS
SI-4102
444-1430EA 1310 EUR InStock
444-1430
MultiPlate Genie® Microplate Shaker
Agitateurs Agitateurs pour microplaques
Fully programmable microplate shaker for four plates (8 with optional tray).
  • Provides consistent well-to-well and plate-to-plate mixing for 96 and 384-well microplates, deep well plates, or PCR plates
  • Will accept PCR Work up racks with standard microplate footprint for use with PCR tubes and strips
  • Featuring programmable speed, time, pulsing/interval mixing - for accurate and repeatable results
Digital true vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray). The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates. Featuring programable speed, time and continuous or pulse time, it’s ideal for achieving accurate and repeatable results.
The Pulse feature allows rapid pulsing or timed ''on/off'' mixing for interval mixing. Ideal for use in PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay applications.
Informations de commande: Optional stacking tray accessory for additional 4 microplates (giving total of 8) is available.
Information de livraison: Supplied with tray for 4 microplates.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité