44505.K2
44505.K2EA
126
EUR
InStock
44505.K2
Tétrahydrofuranne ≥99.8% non stabilisé, Biograde pour la synthèse d'ADN et de peptides
Tétrahydrofuranne
Purity: 99.8%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Nucleic acid and peptide synthesis