44585.WY
44585.WYEA
90.8
EUR
InStock
44585.WY
Nichrome solution d'attaque
Nichrome
Nichrome Etchant is a high purity system based on ceric ammonium nitrate. It provides clean, precise etching of evaporated nichrome layers for the preparation of thin film circuits. This product can be operated at room temperature or elevated temperature to increase etch rate.
Semiconductor-grade ingredients for ultra-high purity
Etchant filter to 0,2 microns to remove particles