44600.36
44600.36EA
235
EUR
InStock
44600.36
Invar®, iron nickel ≥99.9% (base métaux), en poudre -325 mesh
Invar®, iron nickel
Purity: 99.9%
Molecular Formula: Fe:Ni 65:35 wt%
Physical Form: Powder
Appearance: Grey
Odour: Odourless
UN No.: UN3089
MDL No.: MFCD01741928