Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.025%
Packaged under argon in resealable ChemSeal™ bottles.
44608.M1
44608.M1EA
342
EUR
InStock
44608.M1
Tétrahydrofuranne, anhydre ≥99.8% stabilisé
Tétrahydrofuranne
Purity: 99.8+%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356