44661.14
44661.14EA
87
EUR
InStock
44661.14
Cobalt oxyde ≥99% (base métaux), nanopoudre
Cobalt oxyde
Purity: 99%
CAS No.: 1308-06-1
Molecular Formula: Co3O4
Formula Weight: 240.80
Physical Form: 50-80nm APS Powder, S.A. 10m2/g
Density: 6.11
Appearance: Black
Melting Point: 900° dec.
Merck Reference: 14,2429
UN No.: 3077
MDL No.: MFCD00010939