44781.14
44781.14EA
82.6
EUR
InStock
44781.14
Gel de silice amorphe ≥99.9% (base métaux), nanopoudre
Silicon(IV) oxide, amorphous
Purity: 99.9%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: 80nm APS Powder
Density: 2.4
Appearance: White
Odour: Odourless
pH: 6
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232