Synonyms:
Zirconium dioxide, Zirconium Oxide
44886.14
44886.14EA
92.9
EUR
InStock
44886.14
Dioxyde de zirconium ≥99% (base métaux sauf Hf), nanopoudre
Dioxyde de zirconium
Purity: 99%
CAS No.: 1314-23-4
Molecular Formula: O2Zr
Formula Weight: 123.22
Physical Form: 15-25nm APS Powder, S.A. 20-30 m2/g
Density: 5.89
Appearance: White
Odour: Odourless
Melting Point: ca 2700°
Boiling Point: ca 5000°
Merck Reference: 14,10180
MDL No.: MFCD00011310