44960.14
44960.14EA
60.6
EUR
InStock
44960.14
Cérium (IV) oxyde ≥99.5% (REO, base d'oxyde de terre rare), nanopoudre
Cérium (IV) oxyde
Purity: 99.5% min
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: 15-30nm APS Powder, S.A. 30-50 m2/g
Density: 7.132
Appearance: Pale yellow
Melting Point: 2600°
Merck Reference: 14,1989
MDL No.: MFCD00010933