Brûleur de sécurité, Gasprofi
Fournisseur: WARTEWIG
6.004.000
451-1385EA 578 EUR InStock
451-1385
Brûleur de sécurité, Gasprofi
Appareils chauffants Brûleurs à gaz
The gasprofi 1 SCS is the laboratory gas burners with IR sensor, button function, foot pedal and safety package SCS. Made of stainless steel and guarantee the highest degree of sterility.
  • Various programs are available for all variants
  • Flaming with button function, top-class security
  • Adapts to any work environment
  • Low consumption of gas and electrical power
Extremely compact and permits fatigue-free operation by an extremely low working height. Removable burner head can even be easily disassembled into its individual components for thorough cleaning. The Safety Control System SCS demonstrates state-of-the-art safety technology which constantly analyzes potential hazards and, if necessary, initiates safety measures, such as an interruption of the gas supply.
Suitable for connection to the fully automatic inoculation loop carousel autoloop Pro. Enables connection to all common types of gas via the central gas supply (natural gas, propane/butane) or via various safety adapters for gas cartridges.
Innovative doubleclick IR sensor: burner only starts when the doubleclick IR sensor is activated twice.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité