Hotplate, precision, PZ44
Fournisseur: GESTIGKEIT HARRY
PZ44/230
453-2136EA 3030 EUR InStock
453-2136
Hotplate, precision, PZ44
Appareils chauffants Plaques chauffantes
Cast iron GG15 plate. Robust hot plate designed for continuous operation with automatic control for precise temperatures from 20 to 450 °C, has four adjustable feet for extra stability. The required temperature is set via the digital controller and automatically remains constant. The actual temperature is displayed continuously via easy to read display.
  • Capacity for up to 150 kg load
  • Unit has low voltage socket on right hand side to allow optional accessories such as timeswitch, contact thermometer or temperature fuse cut-out to be connected
The unit can be used in three heating stages: 825, 1650 or 3300 Watt. Each stage can be fine tuned from 10 to 100% by an electronic output controller to prevent overheating and afterheating due to excessive power. The 825 and 3300 W heating stages heat the entire plate area, while the special 1650 W stage heats only the right hand section of the plate. Creeping heat means the temperature drops off from the right-hand section to the left-hand plate edge, by approximately 40%.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: Pour une installation, un IQOQ, un étalonnage accrédité selon ISO17025, un entretien ou une réparation, contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité