45596.K2
45596.K2EA
170
EUR
InStock
45596.K2
Acide sulfurique ≥72% (p/p)
Acide sulfurique
CAS No.: 7664-93-9
Molecular Formula: H2O4S
Formula Weight: 98.08
Physical Form: Liquid
Appearance: Colourless to pale yellow
Odour: Acrid
UN No.: 1830
MDL No.: MFCD00064589