Packaged under argon.
45915.14
45915.14EA
487
EUR
InStock
45915.14
Gadolinium ≥99.9% (REO, base d'oxyde de terre rare), en poudre <200 mesh
Gadolinium
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00011022