46314.14
46314.14EA
38.1
EUR
InStock
46314.14
Bismuth (III) oxyde ≥99.9%, nanopoudre
Bismuth (III) oxyde
Purity: 99.9%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: 80-200nm APS Powder, S.A. 3.8-5.5m2/g
Density: 8.9
Appearance: Yellow
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462