Precision digital thermometer (NTC and Pt100) with app connection, testo 110
Fournisseur: TESTO
0563.0110
471-1335EA 126 EUR InStock
471-1335
Precision digital thermometer (NTC and Pt100) with app connection, testo 110
Thermomètres Thermomètres numériques
The compact temperature measuring instrument testo 110 convinces every day in use, shows its strengths wherever measurements are to be made at particularly low temperatures or where highly precise temperatures are to be determined.
The fast temperature all-rounder can show its strengths particularly in applications in laboratories or in the food environment: The optionally available NTC or Pt100 probes cover a comprehensive range of measuring tasks and ensure exact results with particularly high accuracy. The digital Pt100 probes can also be calibrated without the measuring instrument, so we can easily continue working with a different probe without downtime. Also with the optional TopSafe protective cover, dust-tight and protected against water jets (protection class IP 65).
The temperature measuring instrument is rounded off by the testo Smart App for smartphones and tablets, such as configuration of testo 110, second display, measurement data memory, display of the measured values (table, graphic progression) and documentation.
Certifications: The testo 110 is HACCP-compliant, certified to EN 13485.
Information de livraison: Includes testo 110 - 1-channel temperature measuring instrument NTC/Pt100 with App connection, TUC probe socket and audible alarm, transport bag, calibration protocol and 3× AA batteries.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité