Pharmaceutical refrigerator, MED-600-S
Fournisseur: KIRSCH PHILIPP
10115
471-2539EA 8929 EUR InStock
471-2539
Pharmaceutical refrigerator, MED-600-S
Réfrigérateurs
Thanks to its long drawers this pharmaceutical fridge is ideal for complementing pharmacies’ furniture. The drawers have a transparent bottom so users can see at a glimpse what is inside.
  • 12 height adjustable drawers, individual drawer division with adjustable length and cross dividers
  • Recirculating air-cooling
  • Optimised ventilation plates to minimise interior temperature differences
  • Automatic defrosting; melt water evaporation
  • Easy-to-read LED temperature display; easy accessible control panel with key switch
  • Micro-processor controlled temperature and thawing regulation to maximise temperature stability
  • Minimum/Maximum temperature memory
  • Safety control prevents freezing
  • Optical and acoustical alarm including power failure alarm
  • Lockable door
With RS-485 interface for computer based temperature documentation.
Power consumption: 340 Watt
Temperature range: 2 to 20 °C
Certifications: According to DIN 58345
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité