ampouled under argon
47185.03
47185.03EA
146
EUR
InStock
47185.03
Diiodure de cobalt, ultra sec ≥99.999% (base métaux)
Diiodure de cobalt
Purity: 99.999%
CAS No.: 15238-00-3
Molecular Formula: CoI2
Formula Weight: 312.74
Physical Form: Powder, Ampouled under argon
Sensitivity: Light Sensitive; Hygroscopic
Odour: Odourless
Melting Point: 515° vac.
Boiling Point: 570° vac.
MDL No.: MFCD00016030