Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F188470002
479-1150EA 125 EUR InStock
479-1150
Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Boîtes Boîtes de congélation
Polystyrene box with polystyrene cover.
  • Practical cold storage of perishable samples
  • Capacity: twelve 1,5 ml microcentrifuge tubes (tubes not included)
  • Holds −15 ºC temperature for 1,5 hours, and maintains 0 ºC for 4 hours when removed from a freezer
The Cryo-Safe™ Cold Box safeguards enzymes and thermolabile substances from thermal degradation. This compact unit is made up of a molded polystyrene box with cover. The interior of the unit is filled with a cryogenic compound and then permanently sealed, to ensure the low temperatures needed to safeguard materials in the cold box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité