Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Fournisseur: THERMO MATRIX TECHNOLOGIES
4110
479-1248EA 7220 EUR InStock
479-1248
Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Applicateurs de films Applicateurs de film de scellage
Thermo Scientific™ SuperSealer ensures a proper seal on plates, blocks and storage tube racks.
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
  • Designed to work with Thermo Scientific Matrix SepraSeal mats and CapMats
Information de livraison: Includes automatic cap, tape and SepraSeal mat applicator.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité