Benchmate vial rack, CryoELITE®, WHEATON®
Fournisseur: DWK Life Sciences
W985810
479-2111EA 279 EUR InStock
479-2111
Benchmate vial rack, CryoELITE®, WHEATON®
Portoirs Portoirs pour cryotubes
The CryoELITE® benchmate rack can be used for both freestanding and round bottom vials. The Loctagon™ bottom allows for the one hand removal of freestanding vial caps.
Holds up to 50 cryogenic vials. Manufactured from polypropylene, the rack is easily celaned in an automatic washer or autoclavable at 121 °C for 20 minutes. Non-skid feet offer additional stability for bench work. Each well has an inner diameter of 12,5 mm and the rack has an overall footprint of 190×100×22 mm. CryoELITE® benchmate vial rack is stackable. The freestanding vials will lock into the rack.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité