Nunc™ Coloured Closures for CryoTube™
Fournisseur: Thermo Fisher Scientific
363436 336078 336088 336081 336092 336094
391-7638EA 403 EUR InStock
391-7638 479-2476 479-2478 479-2477 479-2479 479-2480
Nunc™ Coloured Closures for CryoTube™
Capuchons Fermetures pour cryotubes
Colour-code your sample vials. Thermo Scientific™ Nunc™ CryoTube™ Coloured caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal. Nunc CryoTubes are the most widely used cryo vials in the world, stemming from the first cryo vials manufactured in the early 1960s. The well-proven design, the high certification level and the large portfolio of vials provide researchers with a choice of the vial best suited for their application and peace of mind that the sample can be retrieved viable and unharmed.
  • These replacement
More Product Information
Limited Time Sale
Certains articles ci-dessous sont en promotion
Collapse All
Désignation Couleur Cdt Numéro de catalogue VWR Quantité
For cryotubes with internal thread, sterile incolore 2.500 479-2476P
Promotion
Valable jusque au 30/06/2025
For cryotubes with internal thread, sterile rouge 2.500 479-2477P
Promotion
Valable jusque au 30/06/2025
For cryotubes with internal thread, sterile bleu 2.500 479-2478P
Promotion
Valable jusque au 30/06/2025
For cryotubes with internal thread, sterile jaune 2.500 479-2479P
Promotion
Valable jusque au 30/06/2025
For cryotubes with internal thread, non sterile incolore 2.500 391-7638P
Promotion
Valable jusque au 30/06/2025
For cryotubes with internal thread, sterile assortiment 2.500 479-2480P
Promotion
Valable jusque au 30/06/2025
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité