Steam sterilisation indicator labels, Comply™ (SteriGage™)
Fournisseur: 3M MEDICAL
M1243A
481-0842EA 899 EUR InStock
481-0842
Steam sterilisation indicator labels, Comply™ (SteriGage™)
Indicateurs de stérilisation
Integrating indicator for use in determining whether the process conditions necessary for sterilisation were met inside each pack. Can be used in all 118 to 138 °C (245 to 280 °F) steam sterilisation cycles.
  • Ink migration allows for easy to read 'accept' or 'reject' results
  • 3M™ Comply™ SteriGage™ Steam Chemical Integrators 1243A, 1243B for pack control monitoring of all 118-138 °C (245-280 °F) steam sterilisation cycles
  • The integrator is designed to correlate with the performance of a BI at 3 time/temperature relationships*
  • Five year shelf life
As an internal pack monitor, the 3M™ Comply™ SteriGage™ Steam Chemical Integrator offers an immediate, accurate and easy to read method of monitoring the sterilisation process conditions inside each pack. It can be used in all steam sterilisers.
Certifications: Confirmed by British Standards Institution (BSI) Kitemark License for compliance with ANSI/AAMI/ISO 11140-1:2014 Type 5 (Category 5) Integrating Indicators
Attention: *The stated values compare with a 1 x 106 spore population of Geobacillus stearothermophilus with a D-value of 1.6 minutes at 121˚C and a z-value of 10.0 ˚C.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité