HPLC isocratic systems diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Fournisseur: Merck
48270-U
48270-UEA 52.6 EUR InStock
48270-U
HPLC isocratic systems diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Étalons Étalons de chromatographie gazeuse
This formulation is designed for evaluating how reliably a chromatographic system is providing such fundamentally important parameters as flow rate, proportioning, and mixing.
Analytes:
Butylparaben 25 μg/ml; Ethyl 4-hydroxybenzoate 14 μg/ml; Methyl 4-hydroxybenzoate 10 μg/ml; Propyl 4-hydroxybenzoate 15 μg/ml
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité