Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.

Potassium dihydrogénophosphate, ULTREX®, Ultrapure, J.T.Baker®

Fournisseur: Avantor
Attention
Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

4921-04EA 473.06 EUR InStock
4921-04
Potassium dihydrogénophosphate, ULTREX®, Ultrapure, J.T.Baker®
Potassium dihydrogénophosphate


Formule: KH₂PO₄
MW: 136.09 g/mol
Point d'ébullition: ~100 °C (1013 hPa)
Point de fusion: 253
Densité: 2.34 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011401
Numéro CAS: 7778-77-0
EINECS: 231-913-4
ATTEINDRE: 01-2119490224-41
Merck Index: 13,07744
Commander maintenant
Résultats des tests de spécification
For Laboratory,Research,or Manufacturing Use
Assay (KH₂PO₄) (dried basis), For Information Only
Loss on Drying at 105°C, For Information Only
Particulate Matter, For Information Only
pH of 5% Solution at 25°C, For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only
Non-Metallic Impurities - Fluoride (F), For Information Only
Non-Metallic Impurities - Halide (as Cl), For Information Only
Non-Metallic Impurities - Nitrogen Compounds (as N), For Information Only
Non-Metallic Impurities - Silicon (Si), For Information Only
Non-Metallic Impurities - Sulfate (SO₄), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Niobium (Nb), For Information Only
Metallic Impurities - Silver (Ag), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 400 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 220 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 215 nm, For Information Only
Certificate Provided Reports Actual Lot Analysis

 

COMMANDE


Appel de prix
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 016 385 011
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 016 385 011
Ces articles ne peuvent être ajoutés au Panier. Veuillez contacter votre service client ou envoyer un e-mail à vwr.be@vwr.com
Une documentation supplémentaire peut être nécessaire pour l'achat de cet article. Un représentant de VWR vous contactera si nécessaire.
Ce produit a été bloqué par votre organisation. Contacter votre service d'achat pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Les produits marqués de ce symbole ne seront bientôt plus disponibles - vente jusqu'à épuisement de stock. Des alternatives peuvent être disponibles en recherchant le code article VWR indiqué ci-dessus. Si vous avez besoin d'une assistance supplémentaire, veuillez contacter notre Service Clientèle au 016 385 011.
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
Glass bottle, amber, wide mouth, black T/E cap 100 g 4921-04

Générer des étiquettes code à barres. Format des étiquettes code à barres.: 
Créer une page catalogue sous forme pdf

claquement

Veuillez sélectionner l'article

Entrer les détails

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité

Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.

Pour en savoir plus, consultez notre Politique de Confidentialité..