Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

di-Sodium hydrogénophosphate, anhydre USP, Multi-Compendial HP, J.T.Baker®

Fournisseur: Avantor
Attention

Management of Change (MOC) category = R

4953-01EA 0 EUR InStock
4953-01
di-Sodium hydrogénophosphate, anhydre USP, Multi-Compendial HP, J.T.Baker®
di-Sodium hydrogen phosphate


Formule: Na₂HPO₄
Point de fusion: 250 °C
Densité: 1.53 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00003496
Numéro CAS: 7558-79-4
EINECS: 231-448-7
ATTEINDRE: 01-2119489797-11
Merck Index: 12,08805
Commander maintenant
Résultats des tests de spécification
Meets JPE Specifications
GMP Manufactured Product
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Identification Passes Test
USP - Loss on Drying at 130°C ≤ 5.0 %
USP - Insoluble Substances ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.06 %
USP - Sulfate (SO₄) ≤ 0.2 %
USP - Arsenic (As) ≤ 16 ppm
JPE - Assay (dried basis) ≥ 99.0 %
JPE - Arsenic (As) ≤ 2 ppm
JPE - Carbonate Passes Test
JPE - Chloride (Cl) ≤ 0.011 %
JPE - Clarity and Color of Solution Passes Test
JPE - Heavy Metals ≤ 10 ppm
JPE - pH (1-100) 9.0 - 9.4
JPE - Sulfate (SO₄) ≤ 0.038 %
JPE - Loss on Drying ≤ 2.0 %
EP - Assay (Na₂HPO₄) (dried basis) 98.0 - 101.0 %
EP - Appearance of Solution Passes Test
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Identification C Passes Test
EP - Identification D Passes Test
EP - Reducing Substances Passes Test
EP - Chloride (Cl) ≤ 200 ppm
EP - Iron (Fe) ≤ 20 ppm
EP - Loss on Drying ≤ 1.0 %
EP - Monosodium Phosphate ≤ 2.5 %
EP - Sulfate (SO₄) ≤ 500 ppm
Endotoxin Concentration (EU/g) ≤ 2.5
Microbial Testing – Total Aerobic Microbial Count (USP) ≤ 10 cfu/g
Microbial Testing – Total Yeast and Mold Count (USP) ≤ 10 cfu/g
Microbial Testing - Salmonella (USP) Negative
Microbial Testing - Escherichia coli (USP) Negative
Microbial Testing - Staphylococcus aureus (USP) Negative
Microbial Testing - Pseudomonas aeruginosa (USP) Negative
Microbial Testing -Bile tolerant gram-negative bacteria Negative
Microbial Testing – Clostridia Negative
Microbial Testing - Candida albicans Negative
Trace Impurities - Cadmium (Cd) ≤ 200 ppb
Trace Impurities - Lead (Pb) ≤ 500 ppb
Trace Impurities - Arsenic (As) ≤ 1500 ppb
Trace Impurities - Mercury (Hg) ≤ 300 ppb
Trace Impurities - Cobalt (Co) ≤ 500 ppb
Trace Impurities - Vanadium (V) ≤ 1000 ppb
Trace Impurities - Nickel (Ni) ≤ 2000 ppb
Trace Impurities - Thallium (Tl) ≤ 800 ppb
Trace Impurities - Gold (Au) ≤ 10000 ppb
Trace Impurities - Palladium (Pd) ≤ 1000 ppb
Trace Impurities - Iridium (Ir) ≤ 1000 ppb
Trace Impurities - Osmium (Os) ≤ 1000 ppb
Trace Impurities - Rhodium (Rh) ≤ 1000 ppb
Trace Impurities - Ruthenium (Ru) ≤ 1000 ppb
Trace Impurities - Selenium (Se) ≤ 8000 ppb
Trace Impurities - Silver (Ag) ≤ 1000 ppb
Trace Impurities - Platinum (Pt) ≤ 1000 ppb
Trace Impurities - Lithium (Li) ≤ 25000 ppb
Trace Impurities - Antimony (Sb) ≤ 9000 ppb
Trace Impurities - Barium (Ba) ≤ 70000 ppb
Trace Impurities - Molybdenum (Mo) ≤ 150000 ppb
Trace Impurities - Copper (Cu) ≤ 30000 ppb
Trace Impurities - Tin (Sn) ≤ 60000 ppb
Trace Impurities - Chromium (Cr) ≤ 110000 ppb
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

COMMANDE

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité