Distributeur à membrane pour contrôle microbiologique, Membrane-Butler, Whatman™
Fournisseur: Whatman products (Cytiva)
10477100
511-5500EA 1560 EUR InStock
511-5500
Distributeur à membrane pour contrôle microbiologique, Membrane-Butler, Whatman™
Membrane Dispensers
Manual butler for dispensing filtration membranes.
  • High reliability
  • Cross contamination risks are minimized
  • Membrane dispensed rapidly
  • Highly suitable for use on sterile benches
  • Compact dimensions for portable use
The Membrane-Butler dispenses individual sterile MicroPlus and ME membrane filters (type 'STL'). The sterile packaging is inserted into the roller system and the system is ready to be used.
Membrane filters for microbiological checks must be handled carefully to ensure that they remain sterile and that quantitative results are being obtained.
The Membrane-Butler offers optimal handling for all MicroPlus and ME membrane filters with the type name ‘STL’. The dispenser box is placed in the Membrane-Butler, the sterile packaging is inserted into the roller system, and the system is ready. With each turn (Manual Butler), a membrane filter is ejected from its sterile packing and can be removed easily with forceps.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité